2,2-Dimethylcyclohexanone ≥95%

Supplier: Matrix Scientific

075715-1G 075715-5G
102542-670EA 384.62 USD
102542-670 102542-672
2,2-Dimethylcyclohexanone ≥95%
2,2-Dimethylcyclohexanone

Hazard: Irritant

Formula: C₈H₁₄O
MW: 126.1962 g/mol
MDL Number: MFCD00134382
CAS Number: 1193-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR