Boc-Ser(Tos)-OMe ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-Methyl-2-((tert-butoxycarbonyl)amino)-3-(tosyloxy)propanoate, Boc-Ser(Tos)-OCH3, (S)-Methyl 2-((tert-butoxycarbonyl)amino)-3-(tosyloxy)propanoate

075711-1G 075711-5G 075711-10G
102542-652EA 49.23 USD
102542-652 102542-654 102542-656
Boc-Ser(Tos)-OMe ≥95%
Boc-Ser(Tos)-OMe

Hazard: Irritant

Formula: C₁₆H₂₃NO₇S
MW: 373.42 g/mol
MDL Number: MFCD01632224
CAS Number: 56926-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR