Etomidate ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-(+)-1-(1-Phenylethyl)-1H-imidazole-5-carboxylic acid ethyl ester, Ethyl (R)-(+)-1-(1-Phenylethyl)-1H-imidazole-5-carboxylate

075710-1G 075710-5G
102542-648EA 321.54 USD
102542-648 102542-650
Etomidate ≥95%
Etomidate

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₂
MDL Number: MFCD00869295
CAS Number: 33125-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR