N-Boc-O-methyl-L-threonine ≥95%

Supplier: Matrix Scientific

Synonyms: (2S,3R)-2-(Boc-amino)-3-methoxybutyric acid, (2S,3R)-2-((tert-Butoxycarbonyl)amino)-3-methoxybutanoic acid, Boc-Thr(Me)-OH

075709-1G 075709-5G
102542-644EA 233.85 USD
102542-644 102542-646
N-Boc-O-methyl-L-threonine ≥95%
N-Boc-O-methyl-L-threonine

Hazard: Irritant

Formula: C₁₀H₁₉NO₅
MW: 233.3 g/mol
MDL Number: MFCD00076990
CAS Number: 48068-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR