4-(4-Hydroxyphenyl)benzonitrile ≥95%

Supplier: Matrix Scientific

075708-10G 075708-100G
102542-640EA 1443.08 USD
102542-640 102542-642
4-(4-Hydroxyphenyl)benzonitrile ≥95%
4-(4-Hydroxyphenyl)benzonitrile

Hazard: Irritant

Formula: C₁₃H₉NO
MDL Number: MFCD00059625
CAS Number: 19812-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR