Benzyl (S)-(+)-glycidyl ether ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-2-((Benzyloxy)methyl)oxirane

075704-100G 075704-25G
102542-622EA 444.62 USD
102542-622 102833-862
Benzyl (S)-(+)-glycidyl ether ≥95%
Benzyl (S)-(+)-glycidyl ether

Hazard: Irritant

Formula: C₁₀H₁₂O₂
Density: 1.072
MDL Number: MFCD00054428
CAS Number: 16495-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR