3-((4-Nitrobenzyl)oxy)-3-oxopropanoic acid ≥95%

Supplier: Matrix Scientific

075703-5G 075703-10G 075703-100G
102542-614EA 20 USD
102542-614 102542-616 102542-618
3-((4-Nitrobenzyl)oxy)-3-oxopropanoic acid ≥95%
3-((4-Nitrobenzyl)oxy)-3-oxopropanoic acid

Hazard: Irritant

Formula: C₁₀H₉NO₆
MDL Number: MFCD00191556
CAS Number: 77359-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR