Ritalinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Phenyl-2-(2-piperidyl)acetic acid

075702-1G 075702-5G
102542-610EA 64.62 USD
102542-610 102542-612
Ritalinic acid ≥95%
Ritalinic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₂
MDL Number: MFCD06200695
CAS Number: 19395-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR