2,4,6-Trimethoxybenzaldehyde ≥95%

Supplier: Matrix Scientific

075701-10G 075701-100G
102542-606EA 126.15 USD
102542-606 102542-608
2,4,6-Trimethoxybenzaldehyde ≥95%
2,4,6-Trimethoxybenzaldehyde

Hazard: Irritant

Formula: C₁₀H₁₂O₄
MDL Number: MFCD00003313
CAS Number: 830-79-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR