(S)-N,N-Dimethyl-3-(naphthalen-1-yloxy)-1-phenylpropan-1-amine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Dapoxetine HCl, (S)-N,N-Dimethyl-alpha-[2-(1-naphthyloxy)ethyl]benzylamine hydrochloride, Dapoxetine hydrochloride, (S)-N,N-Dimethyl-3-(1-naphthyloxy)-1-phenylpropylamine hydrochloride

075698-1G 075698-5G
102542-594EA 533.85 USD
102542-594 102542-596
(S)-N,N-Dimethyl-3-(naphthalen-1-yloxy)-1-phenylpropan-1-amine hydrochloride ≥95%
(S)-N,N-Dimethyl-3-(naphthalen-1-yloxy)-1-phenylpropan-1-amine hydrochloride

Hazard: Irritant

Formula: C₂₁H₂₄ClNO
MW: 341.88 g/mol
MDL Number: MFCD08272809
CAS Number: 129938-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR