Methyl-5-fluoro-2-methylbenzoate ≥95%

Supplier: Matrix Scientific

075697-1G 075697-5G 075697-10G
102542-588EA 107.69 USD
102542-588 102542-590 102542-592
Methyl-5-fluoro-2-methylbenzoate ≥95%
Methyl 5-fluoro-2-methylbenzoate

Hazard: Irritant

Formula: C₉H₉FO₂
MDL Number: MFCD00221457
CAS Number: 175278-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR