4-Chlorobenzamidine hydrochloride ≥95%

Supplier: Matrix Scientific

075696-1G 075696-5G 075696-10G
102542-582EA 90.77 USD
102542-582 102542-584 102542-586
4-Chlorobenzamidine hydrochloride ≥95%
4-Chlorobenzamidine hydrochloride

Hazard: Irritant

Formula: C₇H₈Cl₂N₂
MDL Number: MFCD00126401
CAS Number: 14401-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR