Methyl-5-bromo-3-(trifluoromethyl)picolinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-bromo-3-(trifluoromethyl)picolinate, Methyl 5-bromo-3-(trifluoromethyl)-2-pyridinecarboxylate

075695-250MG 075695-1G
102542-578EA 461.54 USD
102542-578 102542-580
Methyl-5-bromo-3-(trifluoromethyl)picolinate ≥95%
Methyl-5-bromo-3-(trifluoromethyl)picolinate

Hazard: Irritant

Formula: C₈H₅BrF₃NO₂
MW: 284.0300 g/mol
MDL Number: MFCD14698001
CAS Number: 1214328-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR