7-Bromobenzofuran-3(2H)-one ≥95%

Supplier: Matrix Scientific

075694-250MG 075694-1G 075694-5G
102542-572EA 456.92 USD
102542-572 102542-574 102542-576
7-Bromobenzofuran-3(2H)-one ≥95%
7-Bromobenzofuran-3(2H)-one

Hazard: Irritant

Formula: C₈H₅BrO₂
MW: 213.0300 g/mol
MDL Number: MFCD03160589
CAS Number: 519018-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR