Naphthalen-2-ylmethanamine hydrochloride ≥95%

Supplier: Matrix Scientific

075693-1G 075693-5G 075693-10G
102542-566EA 169.23 USD
102542-566 102542-568 102542-570
Naphthalen-2-ylmethanamine hydrochloride ≥95%
Naphthalen-2-ylmethanamine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₂ClN
MW: 193.6700 g/mol
MDL Number: MFCD00137202
CAS Number: 2241-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR