2,5-Dichlorothiophene-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

075690-250MG 075690-1G 075690-5G
102542-550EA 427.69 USD
102542-550 102542-552 102542-554
2,5-Dichlorothiophene-3-carbaldehyde ≥95%
2,5-Dichlorothiophene-3-carbaldehyde

Hazard: Irritant

Formula: C₅H₂Cl₂OS
MDL Number: MFCD01571097
CAS Number: 61200-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR