(3-Fluoropyridin-4-yl)methanamine ≥95%

Supplier: Matrix Scientific

075689-250MG 075689-1G 075689-5G
102542-544EA 350.81 USD
102542-544 102542-546 102542-548
(3-Fluoropyridin-4-yl)methanamine ≥95%
(3-Fluoropyridin-4-yl)methanamine

Hazard: Irritant

Formula: C₆H₇FN₂
MW: 126.1300 g/mol
MDL Number: MFCD11100727
CAS Number: 870063-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR