Paroxetine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: (3S,4R)-3-((Benzo[d][1,3]dioxol-5-yloxy)methyl)-4-(4-fluorophenyl)piperidine hydrochloride

075688-1G 075688-5G
102542-540EA 141.54 USD
102542-540 102542-542
Paroxetine hydrochloride ≥95%
Paroxetine hydrochloride

Hazard: Irritant

Formula: C₁₉H₂₁ClFNO₃
CAS Number: 78246-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR