6-Bromo-4-(trifluoromethyl)quinolin-2(1H)-one ≥95%

Supplier: Matrix Scientific

075687-250MG 075687-1G 075687-5G
102542-534EA 355.38 USD
102542-534 102542-536 102542-538
6-Bromo-4-(trifluoromethyl)quinolin-2(1H)-one ≥95%
6-Bromo-4-(trifluoromethyl)quinolin-2(1H)-one

Hazard: Irritant

Formula: C₁₀H₅BrF₃NO
MDL Number: MFCD11846359
CAS Number: 328955-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR