7-Methoxynaphthalen-1-ol ≥95%

Supplier: Matrix Scientific

075686-250MG 075686-1G 075686-5G
102542-528EA 456.92 USD
102542-528 102542-530 102542-532
7-Methoxynaphthalen-1-ol ≥95%
7-Methoxynaphthalen-1-ol

Hazard: Irritant

Formula: C₁₁H₁₀O₂
MDL Number: MFCD01314215
CAS Number: 67247-13-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR