1-Ethyl-7-nitro-1,2,3,4-tetrahydroquinoline ≥95%
Supplier: Matrix Scientific
075685-1G
075685-5G
075685-10G
102542-522EA
90.77
USD
102542-522
102542-524
102542-526
1-Ethyl-7-nitro-1,2,3,4-tetrahydroquinoline ≥95%
1-Ethyl-7-nitro-1,2,3,4-tetrahydroquinoline
Hazard: Irritant
Formula:
C₁₁H₁₄N₂O₂ MW: 206.2400 g/mol |
MDL Number:
MFCD07368631 CAS Number: 57883-28-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...