1H-Pyrrolo[3,2-c]pyridine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

075680-250MG 075680-1G 075680-5G
102542-494EA 738.46 USD
102542-494 102542-496 102542-498
1H-Pyrrolo[3,2-c]pyridine-2-carboxylic acid ≥95%
1H-Pyrrolo[3,2-c]pyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₈H₆N₂O₂
MW: 162.1500 g/mol
MDL Number: MFCD09743299
CAS Number: 800401-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR