(S)-tert-Butyl-3-hydroxybutanoate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl (S)-(+)-3-hydroxybutyrate

075679-1G 075679-5G
102542-490EA 355.38 USD
102542-490 102542-492
(S)-tert-Butyl-3-hydroxybutanoate ≥95%
(S)-tert-Butyl-3-hydroxybutanoate

Hazard: Irritant

Formula: C₈H₁₆O₃
MW: 160.21 g/mol
MDL Number: MFCD00040559
CAS Number: 82578-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR