4-Bromo-2,6-dimethylbenzoic acid ≥95%

Supplier: Matrix Scientific

075678-1G 075678-5G
102542-486EA 347.69 USD
102542-486 102542-488
4-Bromo-2,6-dimethylbenzoic acid ≥95%
4-Bromo-2,6-dimethylbenzoic acid

Hazard: Irritant

Formula: C₉H₉BrO₂
MDL Number: MFCD09965997
CAS Number: 74346-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR