2-(Difluoromethoxy)phenylacetic acid ≥95%

Supplier: Matrix Scientific

075677-1G 075677-5G
102542-482EA 427.69 USD
102542-482 102542-484
2-(Difluoromethoxy)phenylacetic acid ≥95%
2-(Difluoromethoxy)phenylacetic acid

Hazard: Irritant

Formula: C₉H₈F₂O₃
MDL Number: MFCD00236237
CAS Number: 86867-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR