Ethyl-6-amino-5-methoxy-1H-indole-2-carboxylate ≥95%

Supplier: Matrix Scientific

075676-250MG 075676-1G 075676-5G
102542-476EA 456.92 USD
102542-476 102542-478 102542-480
Ethyl-6-amino-5-methoxy-1H-indole-2-carboxylate ≥95%
Ethyl-6-amino-5-methoxy-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₃
MW: 234.25 g/mol
MDL Number: MFCD08064224
CAS Number: 107575-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR