Methyl-2-(bromomethyl)-3-chlorobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-(bromomethyl)-3-chlorobenzoate

075675-250MG 075675-1G 075675-5G
102542-470EA 355.38 USD
102542-470 102542-472 102542-474
Methyl-2-(bromomethyl)-3-chlorobenzoate ≥95%
Methyl 2-(bromomethyl)-3-chlorobenzoate

Hazard: Irritant

Formula: C₉H₈BrClO₂
MW: 263.5156 g/mol
MDL Number: MFCD04114322
CAS Number: 188187-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR