3-Methoxybenzylamine ≥95%
Supplier: Matrix Scientific
Synonyms:
m-Anisylamine, 3-(Aminomethyl)anisole
075671-5G
075671-10G
102542-450EA
33.85
USD
102542-450
102542-452
3-Methoxybenzylamine ≥95%
3-Methoxybenzylamine
Hazard: Irritant
Formula:
C₈H₁₁NO MW: 137.18 g/mol |
MDL Number:
MFCD00008115 CAS Number: 5071-96-5 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...