2'-Chloro-5'-fluoroacetophenone ≥95%

Supplier: Matrix Scientific

075669-1G 075669-5G 075669-25G
102542-440EA 49.23 USD
102542-440 102542-442 102833-860
2'-Chloro-5'-fluoroacetophenone ≥95%
2'-Chloro-5'-fluoroacetophenone

Hazard: Irritant

Formula: C₈H₆ClFO
MW: 172.59 g/mol
MDL Number: MFCD00042574
CAS Number: 2965-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR