3-Chloro-5-fluorobenzonitrile ≥95%
Supplier: Matrix Scientific
075667-1G
075667-5G
075667-10G
102542-428EA
72.31
USD
102542-428
102542-430
102542-432
3-Chloro-5-fluorobenzonitrile ≥95%
3-Chloro-5-fluorobenzonitrile
Hazard: Toxic
Formula:
C₇H₃ClFN MW: 155.56 g/mol Melting Pt: 54-59°C |
MDL Number:
MFCD03407967 CAS Number: 327056-73-5 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...