4-Bromo-o-anisidine ≥95%

Supplier: Matrix Scientific

075664-1G 075664-5G
102542-412EA 15.38 USD
102542-412 102542-414
4-Bromo-o-anisidine ≥95%
4-Bromo-o-anisidine

Hazard: Toxic

Formula: C₇H₈BrNO
MDL Number: MFCD01204266
CAS Number: 59557-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR