Ethyl picolinate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-pyridinecarboxylate, Ethyl 2-picolinate
075663-10G
075663-100G
102542-408EA
72.31
USD
102542-408
102542-410
Ethyl picolinate ≥95%
Ethyl picolinate
Hazard: Irritant
Formula:
C₈H₉NO₂ MW: 151.16 g/mol |
MDL Number:
MFCD00006292 CAS Number: 2524-52-9 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...