Warfarin sodium ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(α-Acetonylbenzyl)-4-hydroxycoumarin sodium salt, 4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin

075658-5G 075658-10G 075658-100G
102542-384EA 72.31 USD
102542-384 102542-386 102542-388
Warfarin sodium ≥95%
Warfarin sodium

Hazard: Irritant

Formula: C₁₉H₁₆NaO₄
MDL Number: MFCD00083223
CAS Number: 129-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR