3-Nitrophenol ≥95%

Supplier: Matrix Scientific

075655-10G 075655-100G
102542-370EA 21.54 USD
102542-370 102542-372
3-Nitrophenol ≥95%
3-Nitrophenol

Hazard: Toxic

Formula: C₆H₅NO₃
Boiling Pt: 194°C/70mm
Melting Pt: 93-98°C
Density: 1.49
Flash Pt: 100°C
MDL Number: MFCD00007240
CAS Number: 554-84-7
UN: 1663
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR