(2R)-(-)-2,3-Epoxypropyl toluene-4-sulfonate ≥95%

Supplier: Matrix Scientific

Synonyms: (2R)-(-)-Glycidyl p-toluenesulphonate, (2R)-(-)-Glycidyl tosylate

075654-10G 075654-100G
102542-366EA 813.85 USD
102542-366 102542-368
(2R)-(-)-2,3-Epoxypropyl toluene-4-sulfonate ≥95%
(2R)-(-)-2,3-Epoxypropyl toluene-4-sulphonate

Hazard: Irritant

Formula: C₁₀H₁₂O₄S
MW: 228.2649 g/mol
MDL Number: MFCD00010834
CAS Number: 113826-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR