3-Hydroxy-4-nitrobenzaldehyde ≥95%

Supplier: Matrix Scientific

075653-5G 075653-25G 075653-100G
102542-362EA 66.15 USD
102542-362 102551-990 102551-992
3-Hydroxy-4-nitrobenzaldehyde ≥95%
3-Hydroxy-4-nitrobenzaldehyde

Hazard: Irritant

Formula: C₇H₅NO₄
MDL Number: MFCD00007109
CAS Number: 704-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR