2,5-Dimethylbenzoic acid ≥95%

Supplier: Matrix Scientific

075652-10G 075652-100G 075652-500G
102542-354EA 90.77 USD
102542-354 102542-356 102542-358
2,5-Dimethylbenzoic acid ≥95%
2,5-Dimethylbenzoic acid

Hazard: Irritant

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
MDL Number: MFCD00002482
CAS Number: 610-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR