Ethyl-2-(2-aminothiazol-4-yl)-2-oxoacetate ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(2-Amino-4-thiazolyl)-2-oxoacetic acid ethyl ester, Ethyl 2-(2-Amino-4-thiazolyl)-2-oxoacetate, Ethyl 2-(2-aminothiazol-4-yl)-2-oxoacetate

075650-100G 075650-25G
102542-346EA 189.23 USD
102542-346 102551-988
Ethyl-2-(2-aminothiazol-4-yl)-2-oxoacetate ≥95%
Ethyl-2-(2-aminothiazol-4-yl)-2-oxoacetate

Hazard: Irritant/Refrigerate

Formula: C₇H₉NO₄S
MDL Number: MFCD00010414
CAS Number: 64987-08-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR