(4-Methoxybenzyl)hydrazine ≥95%

Supplier: Matrix Scientific

075649-1G 075649-5G
102542-338EA 461.54 USD
102542-338 102542-340
(4-Methoxybenzyl)hydrazine ≥95%
(4-Methoxybenzyl)hydrazine

Hazard: Irritant

Formula: C₈H₁₂N₂O
MW: 152.2 g/mol
Boiling Pt: 145 °C (12 Torr)
Melting Pt: 34…36 °C
MDL Number: MFCD07786530
CAS Number: 140-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR