1-(4-Fluorophenyl)piperazine dihydrochloride ≥95%

Supplier: Matrix Scientific

075646-10G 075646-100G
102542-322EA 15.38 USD
102542-322 102542-324
1-(4-Fluorophenyl)piperazine dihydrochloride ≥95%
1-(4-Fluorophenyl)piperazine dihydrochloride

Hazard: Toxic

Formula: C₁₀H₁₅Cl₂FN₂
MW: 253.15 g/mol
Melting Pt: ∼ 234 °C (dec.)
MDL Number: MFCD00012765
CAS Number: 64090-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR