6-Methylnicotinamide ≥95%
Supplier: Matrix Scientific
Synonyms:
6-Methylpyridine-3-carboxamide
075643-1G
075643-5G
075643-25G
102542-306EA
73.85
USD
102542-306
102542-308
102551-986
6-Methylnicotinamide ≥95%
6-Methylnicotinamide
Hazard: Irritant
Formula:
C₇H₈N₂O MW: 136.15 g/mol |
MDL Number:
MFCD00006342 CAS Number: 6960-22-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...