3-Amino-4-nitrobenzonitrile ≥95%

Supplier: Matrix Scientific

075642-1G 075642-5G
102542-302EA 569.23 USD
102542-302 102542-304
3-Amino-4-nitrobenzonitrile ≥95%
3-Amino-4-nitrobenzonitrile

Hazard: Irritant

Formula: C₇H₅N₃O₂
MDL Number: MFCD18389919
CAS Number: 99512-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR