(S)-2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: H-Asp(OMe)-OH·HCl, L-Aspartic acid 4-methyl ester hydrochloride, L-Aspartatic Acid 4-Methyl Ester Hydrochloride, L-Aspartic acid β-methyl ester hydrochloride, H-Asp(OMe)-OH.HCl, H-Asp(OMe)-OH β-Methyl L-aspartate hydrochloride

075640-5G 075640-100G
102542-290EA 150.77 USD
102542-290 102542-294
(S)-2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride ≥95%
(S)-2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride

Hazard: Irritant

Formula: C₅H₉NO₄·HCl
MW: 183.6 g/mol
MDL Number: MFCD00038972
CAS Number: 16856-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR