4-(Boc-amino)pyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
4-(Boc-Amino)pyridine
075637-5G
075637-25G
075637-100G
102542-276EA
63.08
USD
102542-276
102551-980
102551-982
4-(Boc-amino)pyridine ≥95%
4-(Boc-amino)pyridine
Hazard: Irritant
Formula:
C₁₀H₁₄N₂O₂ MW: 194.23 g/mol Melting Pt: 147-150°C |
MDL Number:
MFCD02179232 CAS Number: 98400-69-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...