2,4,5-Trichlorophenol ≥95%

Supplier: Matrix Scientific

075636-1G 075636-5G 075636-10G
102542-268EA 33.85 USD
102542-268 102542-270 102542-272
2,4,5-Trichlorophenol ≥95%
2,4,5-Trichlorophenol

Hazard: Irritant

Formula: C₆H₃Cl₃O
MW: 197.45 g/mol
MDL Number: MFCD00002170
CAS Number: 95-95-4
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR