Ethyl-1H-indazole-7-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 1H-indazole-7-carboxylate

075633-250MG 075633-1G 075633-5G
102542-254EA 355.38 USD
102542-254 102542-256 102542-258
Ethyl-1H-indazole-7-carboxylate ≥95%
Ethyl 1H-indazole-7-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂
MW: 190.2000 g/mol
MDL Number: MFCD07371560
CAS Number: 885278-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR