(R)-2-Amino-3-methoxypropanoic acid hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: O-Methyl-D-serine hydrochloride

075632-1G 075632-5G 075632-10G
102542-248EA 215.38 USD
102542-248 102542-250 102542-252
(R)-2-Amino-3-methoxypropanoic acid hydrochloride ≥95%
(R)-2-Amino-3-methoxypropanoic acid hydrochloride

Hazard: Irritant

Formula: C₄H₁₀ClNO₃
MW: 155.58 g/mol
MDL Number: MFCD11099371
CAS Number: 86118-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR