trans-(1R,2R)-N,N'-Bismethyl-1,2-cyclohexanediamine dihydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
trans-(1R,2R)-(-)-N,N'-Dimethyl-1,2-cyclohexanediamine dihydrochloride
075629-1G
075629-5G
102542-234EA
233.85
USD
102542-234
102542-236
trans-(1R,2R)-N,N'-Bismethyl-1,2-cyclohexanediamine dihydrochloride ≥95%
trans-(1R,2R)-N,N'-Bismethyl-1,2-cyclohexanediamine dihydrochloride
Hazard: Irritant
Formula:
C₈H₂₀Cl₂N₂ MW: 215.17 g/mol |
MDL Number:
MFCD09749879 CAS Number: 70708-33-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...