S-Trityl-L-cysteine ≥95%

Supplier: Matrix Scientific

Synonyms: H-Cys(Trt)-OH

075627-5G 075627-10G 075627-100G
102542-222EA 21.54 USD
102542-222 102542-224 102542-226
S-Trityl-L-cysteine ≥95%
S-Trityl-L-cysteine

Hazard: Irritant

Formula: C₂₂H₂₁NO₂S
Melting Pt: 182 °C (dec.)
MDL Number: MFCD00002611
CAS Number: 2799-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR