5-Chlorosalicylaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-2-hydroxybenzaldehyde

075626-10G 075626-100G
102542-218EA 53.85 USD
102542-218 102542-220
5-Chlorosalicylaldehyde ≥95%
5-Chlorosalicylaldehyde

Hazard: Irritant

Formula: C₇H₅ClO₂
MW: 156.57 g/mol
MDL Number: MFCD00003331
CAS Number: 635-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR